
1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:876317-19-0
:
Formula:C₁₀H₁₅NO₅
MDL Number:MFCD02668169
:229.2298 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together