Methyl-2-morpholino-2-phenylacetate hydrochloride
Catalog # APOSOR300892-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.75 g/mol
Storage Temperature:Ambient
CAS Number:27594-62-3
Formula:C₁₃H₁₈ClNO₃
MDL Number:MFCD28099762