4-Methoxybenzamidine hydrochloride 97%
Catalog # APOSOR300727-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:186.64 g/mol
Storage Temperature:Ambient
CAS Number:51721-68-7
Formula:C₈H₁₀N₂O
MDL Number:MFCD00466114