Order Entry
Czech Republic
ContactUsLinkComponent
Boc-Tic-OH 98%
Irritant
Boc-Tic-OH 98%
Catalog # APOSOR30067-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Boc-Tic-OH 98%
Catalog # APOSOR30067-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :277.32 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:78879-20-6
Formula:C₁₅H₁₉NO₄
MDL Number:MFCD00143845