
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146256-98-6
:
Formula:C₁₂H₁₉NO₅
MDL Number:MFCD06656201
:257.2830 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together