3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride
Catalog # APOSOR300517-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.69 g/mol
Storage Temperature:Ambient
CAS Number:13582-92-8
Formula:C₁₁H₁₄ClNO₃
MDL Number:MFCD11227230