
Ethyl-1-tert-butyl-5-methyl-1H-pyrazole-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:519056-54-3
:
Formula:C₁₁H₁₈N₂O₂
MDL Number:MFCD01313625
:210.28 g/mol
Boiling Pt:130 °C/15mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together