8-Chloro[1,2,4]triazolo[4,3-a]pyrazine
Catalog # APOSOR300026-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.56 g/mol
Storage Temperature:Ambient
CAS Number:68774-77-6
Formula:C₅H₃ClN₄
MDL Number:MFCD08272086