Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate
Catalog # APOSOR300025-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.18 g/mol
Storage Temperature:Ambient
CAS Number:723286-67-7
Formula:C₈H₈N₄O₂
MDL Number:MFCD09909674