6-Bromoimidazo[1,2-a]pyrazin-8-amine
Catalog # APOSOR300017-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.0400 g/mol
Storage Temperature:Ambient
CAS Number:117718-84-0
Formula:C₆H₅BrN₄
MDL Number:MFCD00874115