
4-(Dimethoxymethyl)-2-methylpyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175277-33-5
:
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD00085137
:168.19 g/mol
Boiling Pt:119…123 °C/5mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together