
Methyl-2,4-dichlorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35112-28-8
:
Formula:C₈H₆Cl₂O₂
MDL Number:MFCD00085117
:205.04 g/mol
Boiling Pt:239 °C
Density:1.386 g/cm³ (25 °C)
Flash Pt:110 °C
EINECS:252-374-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together