

3-Iodobenzylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3718-88-5
:
Formula:C₇H₉ClIN
MDL Number:MFCD00012857
:269.5105 g/mol
Melting Pt:188…190 °C
EINECS:223-068-5
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together