
5-Methoxy-1,3-benzothiazol-2-amine 98%
Catalog # APOSOR2965-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:180.23 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:54346-87-1
Formula:C₈H₈N₂OS
MDL Number:MFCD00464314