
(2Z)-3-Hydroxy-2-phenylacrylonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22252-92-2
:
Formula:C₉H₇NO
MDL Number:MFCD00137446
:145.16 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together