

3,4-Dichlorophenyl isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6590-94-9
:
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00037828
:204.08 g/mol
Boiling Pt:134…136 °C/7mmHg
Density:1.422 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:229-526-0
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Frequently Bought Together