
1-(3-aminopropyl)pyrrolidin-2-one
Catalog # APOSOR2880T-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.202 g/mol
Boiling Pt:120…123 °C/1mmHg
Density:1.014 g/cm³ (25 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:231-632-7
UN:3267
ADR:8,II
:GHS05
CAS Number:7663-77-6
Formula:C₇H₁₄N₂O
MDL Number:MFCD00003201