
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1585-17-7
:
Formula:C₁₁H₁₄Cl₂
MDL Number:MFCD00013681
:217.14 g/mol
Boiling Pt:180 °C/20mmHg
Melting Pt:105…107 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together