Order Entry
Czech Republic
ContactUsLinkComponent
Sodium-DL-3-hydroxybutyrate 98%
Irritant
Sodium-DL-3-hydroxybutyrate 98%
Catalog # APOSOR28800-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Sodium-DL-3-hydroxybutyrate 98%
Catalog # APOSOR28800-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :126.09 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:150-83-4
Formula:C₄H₈O₃
MDL Number:MFCD00016716