Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-3-hydroxypyridine 98%
Irritant
2-Chloro-3-hydroxypyridine 98%
Catalog # APOSOR28674-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Chloro-3-hydroxypyridine 98%
Catalog # APOSOR28674-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :129.55 g/mol
Melting Pt:168…172 °C
EINECS:229-635-3
 :GHS07
Storage Temperature:Ambient
CAS Number:6636-78-8
Formula:C₅H₄ClNO
MDL Number:MFCD00006235