
Methyl-N-methylanthranilate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85-91-6
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00017183
:165.19 g/mol
Boiling Pt:256…258 °C
Melting Pt:17…19 °C
Density:1.126 g/cm³ (25 °C)
Flash Pt:113 °C
EINECS:201-642-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together