
Isopropyltriphenylphosphonium bromide
Catalog # APOSOR28315-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:385.2772 g/mol
Melting Pt:233…236 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1530-33-2
Formula:C₂₁H₂₂BrP
MDL Number:MFCD00064807