Order Entry
Czech Republic
ContactUsLinkComponent
Dimethyl-cis-cyclohexane-1,2-dicarboxylate 96%
Irritant
Dimethyl-cis-cyclohexane-1,2-dicarboxylate 96%
Catalog # APOSOR28288-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Dimethyl-cis-cyclohexane-1,2-dicarboxylate 96%
Catalog # APOSOR28288-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.2300 g/mol
Boiling Pt:111 °C
Density:1.12 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:1687-29-2
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00043768