
1-(Chloromethyl)cyclohexanecarbonitrile
Catalog # APOSOR27708-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.64 g/mol
Boiling Pt:83…86 °C/0.2mmHg
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
CAS Number:112905-95-0
Formula:C₈H₁₂ClN
MDL Number:MFCD01570824