Order Entry
Czech Republic
ContactUsLinkComponent
(4-Methoxybenzyl)triphenylphosphonium bromide
Irritant
(4-Methoxybenzyl)triphenylphosphonium bromide
Catalog # APOSOR27696-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(4-Methoxybenzyl)triphenylphosphonium bromide
Catalog # APOSOR27696-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:1530-38-7
Formula:C₂₆H₂₄BrOP