
1-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5334-99-6
:
Formula:C₆H₇N₅
MDL Number:MFCD00128302
Melting Pt:268…270 °C
:149.16 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together