
2-(4-Methoxyphenoxy)-1-ethanamine, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50800-92-5
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00052978
:167.21 g/mol
Boiling Pt:127…128 °C/2mmHg
Melting Pt:38…40 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together