Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
2,6-Dichloro-4-methylnicotinamide
Catalog # APOSOR27097-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dichloro-4-methylnicotinamide
Catalog # APOSOR27097-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.04 g/mol
Boiling Pt:129…132 °C/7mmHg
 :GHS07
Storage Temperature:Ambient
CAS Number:38841-54-2
Formula:C₇H₆Cl₂N₂O
MDL Number:MFCD00052636