
5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile
Catalog # APOSOR27087-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.61 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:23819-92-3
Formula:C₈H₇ClN₂O
MDL Number:MFCD00052652