
Ethyl-2-bromobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6091-64-1
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00015443
:229.07 g/mol
Boiling Pt:255 °C
Density:1.443 g/cm³ (25 °C)
Flash Pt:>113 °C
EINECS:228-034-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together