Order Entry
Czech Republic
ContactUsLinkComponent
L-Valine ethyl ester hydrochloride 98%
L-Valine ethyl ester hydrochloride 98%
Catalog # APOSOR26830-100G
Supplier:  Apollo Scientific
CAS Number:  
L-Valine ethyl ester hydrochloride 98%
Catalog # APOSOR26830-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :181.66 g/mol
Melting Pt:102…105 °C
Storage Temperature:Ambient
CAS Number:17609-47-1
Formula:C₇H₁₆ClNO₂
MDL Number:MFCD00012511