

3,4-Dimethoxy-3-cyclobutene-1,2-dione 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5222-73-1
:
Formula:C₆H₆O₄
MDL Number:MFCD00101316
:142.11 g/mol
Melting Pt:55…57 °C
:GHS07
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together