
4'-Methoxy-3'-nitroacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6277-38-9
:
Formula:C₉H₉NO₄
MDL Number:MFCD00017023
:195.18 g/mol
Melting Pt:97…100 °C
EINECS:228-476-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together