Order Entry
Czech Republic
ContactUsLinkComponent
5-Methoxy-2-nitrophenol 97%
Irritant
5-Methoxy-2-nitrophenol 97%
Catalog # APOSOR26237-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methoxy-2-nitrophenol 97%
Catalog # APOSOR26237-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :169.14 g/mol
Melting Pt:92…94 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:704-14-3
Formula:C₇H₇NO₄
MDL Number:MFCD00100932