Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(5-Methyl-1,3-thiazol-2-yl)methanol
(5-Methyl-1,3-thiazol-2-yl)methanol
(5-Methyl-1,3-thiazol-2-yl)methanol
APOSOR2604-250MG
:
Apollo Scientific
:
(5-Methyl-1,3-thiazol-2-yl)methanol
APOSOR2604-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
129.18 g/mol
:
GHS07
:
Refrigerator
:
202932-04-5
:
C₅H₇NOS
:
MFCD08060774