
3-(2-Methylpyrimidin-4-yl)benzenesulphonyl chloride
Catalog # APOSOR25988-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:268.72 g/mol
Melting Pt:174…175 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:465514-07-2
Formula:C₁₁H₉ClN₂O₂S
MDL Number:MFCD03659718