Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethylchroman-6-sulphonyl chloride 95%
Corrosive
2,2-Dimethylchroman-6-sulphonyl chloride 95%
Catalog # APOSOR25965-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2,2-Dimethylchroman-6-sulphonyl chloride 95%
Catalog # APOSOR25965-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :260.74 g/mol
Melting Pt:92…94 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:131880-55-2
Formula:C₁₁H₁₃ClO₃S
MDL Number:MFCD03407324