Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1H-Pyrazol-1-yl)aniline
2-(1H-Pyrazol-1-yl)aniline
2-(1H-Pyrazol-1-yl)aniline
APOSOR25934-250MG
:
Apollo Scientific
:
2-(1H-Pyrazol-1-yl)aniline
APOSOR25934-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
159.1879 g/mol
:
GHS07
:
Ambient
:
54705-91-8
:
C₉H₉N₃
:
MFCD03086227