Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethyl-N-pyridin-4-ylpropionamide
2,2-Dimethyl-N-pyridin-4-ylpropionamide
Catalog # APOSOR25910-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethyl-N-pyridin-4-ylpropionamide
Catalog # APOSOR25910-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.24 g/mol
Storage Temperature:Ambient
CAS Number:70298-89-4
Formula:C₁₀H₁₄N₂O
MDL Number:MFCD00996248