
Tris(2-methylphenyl)phosphine 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6163-58-2
:
Formula:C₂₁H₂₁P
MDL Number:MFCD00008514
:304.37 g/mol
Melting Pt:123…129 °C
EINECS:228-193-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together