Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-5-bromo-2-furoate 97%
Methyl-5-bromo-2-furoate 97%
  APOSOR25651-25G
 :  
Methyl-5-bromo-2-furoate 97%
  APOSOR25651-25G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

 :
 :205.01 g/mol
 :Ambient
 :2527-99-3
 :C₆H₅BrO₃
 :MFCD00092313