
4,5,6,7-Tetrahydro-1,3-benzothiazol-2-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2933-29-1
:
Formula:C₇H₁₀N₂S
MDL Number:MFCD00022851
:154.24 g/mol
Melting Pt:86…90 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together