
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
Catalog # APOSOR25017-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.18 g/mol
Melting Pt:252…254 °C
:GHS07
Storage Temperature:Ambient
CAS Number:98488-10-9
Formula:C₈H₉N₃O
MDL Number:MFCD00210153