
ω-Bromo-4-chloro-3-nitroacetophenone, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22019-49-4
:
Formula:C₈H₅BrClNO₃
Melting Pt:159…161 °C
Storage Temperature:Refrigerator
ADR:8,III
:278.49
UN:3261
:GHS05
Frequently Bought Together