Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methylthiazole-2-carbaldehyde
5-Methylthiazole-2-carbaldehyde
5-Methylthiazole-2-carbaldehyde
APOSOR2494-1G
:
Apollo Scientific
:
5-Methylthiazole-2-carbaldehyde
APOSOR2494-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
127.1643 g/mol
:
80…81 °C/7.5mmHg
:
GHS07
:
Ambient
:
13838-78-3
:
C5H5NOS
:
MFCD08060733