
3-Benzothiazol-2-ylpropionic acid
Catalog # APOSOR2471-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.25 g/mol
Melting Pt:108…110 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:29198-86-5
Formula:C₁₀H₉NO₂S
MDL Number:MFCD01764690