

N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride
Catalog # APOSOR24559-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:268.62 g/mol
Melting Pt:134…136 °C
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:10429-82-0
Formula:C₁₁H₁₆Cl₃N
MDL Number:MFCD00185654