Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
Irritant
4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
Catalog # APOSOR24331-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
Catalog # APOSOR24331-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.65 g/mol
Melting Pt:122…125 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:175137-21-0
Formula:C₇H₅ClN₂S
MDL Number:MFCD00205202