
3-Nitro-1H-pyrazole
Catalog # APOSOR24208-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:113.08 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
CAS Number:26621-44-3
Formula:C₃H₃N₃O₂
MDL Number:MFCD00238787