
3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:644-34-8
:
Formula:C₁₆H₁₂O₃
MDL Number:MFCD00022990
Melting Pt:123 °C
:252.27 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together